Merck
  • High fat diet induces microbiota-dependent silencing of enteroendocrine cells.
High fat diet induces microbiota-dependent silencing of enteroendocrine cells.
eLife (2019-12-04)
Lihua Ye, Olaf Mueller, Jennifer Bagwell, Michel Bagnat, Rodger A Liddle, John F Rawls
ABSTRACT
Enteroendocrine cells (EECs) are specialized sensory cells in the intestinal epithelium that sense and transduce nutrient information. Consumption of dietary fat contributes to metabolic disorders, but EEC adaptations to high fat feeding were unknown. Here, we established a new experimental system to directly investigate EEC activity in vivo using a zebrafish reporter of EEC calcium signaling. Our results reveal that high fat feeding alters EEC morphology and converts them into a nutrient insensitive state that is coupled to endoplasmic reticulum (ER) stress. We called this novel adaptation 'EEC silencing'. Gnotobiotic studies revealed that germ-free zebrafish are resistant to high fat diet induced EEC silencing. High fat feeding altered gut microbiota composition including enrichment of Acinetobacter bacteria, and we identified an Acinetobacter strain sufficient to induce EEC silencing. These results establish a new mechanism by which dietary fat and gut microbiota modulate EEC nutrient sensing and signaling.
Sigma-Aldrich
Nω-Nitro-L-arginine methyl ester hydrochloride, ≥97% (TLC), powder
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)
Sigma-Aldrich
Phloridzin dihydrate, from apple wood, ≥99% (HPLC)
Sigma-Aldrich
Brefeldin A, BioXtra, ≥99% (HPLC and TLC), Molecular Biology
Sigma-Aldrich
Peroxide Assay Kit, sufficient for 250 colorimetric tests
Sigma-Aldrich
Gentamicin sulfate salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Thapsigargin, ≥98% (HPLC), solid film
Sigma-Aldrich
Orlistat, ≥98%, solid
Millipore
Millex-SV Filter Unit (Sterile), pore size 5.0 μm, diam. 25 mm, PVDF membrane, hydrophilic, medical device