Merck
  • Immunopurification of Acetylcholinesterase from Red Blood Cells for Detection of Nerve Agent Exposure.
Immunopurification of Acetylcholinesterase from Red Blood Cells for Detection of Nerve Agent Exposure.
Chemical research in toxicology (2017-09-12)
Alicia J Dafferner, Lawrence M Schopfer, Gaoping Xiao, John R Cashman, Udaya Yerramalla, Rudolph C Johnson, Thomas A Blake, Oksana Lockridge
ABSTRACT
Nerve agents and organophosphorus pesticides make a covalent bond with the active site serine of acetylcholinesterase (AChE), resulting in inhibition of AChE activity and toxic symptoms. AChE in red blood cells (RBCs) serves as a surrogate for AChE in the nervous system. Mass spectrometry analysis of adducts on RBC AChE could provide evidence of exposure. Our goal was to develop a method of immunopurifying human RBC AChE in quantities adequate for detecting exposure by mass spectrometry. For this purpose, we immobilized 3 commercially available anti-human acetylcholinesterase monoclonal antibodies (AE-1, AE-2, and HR2) plus 3 new monoclonal antibodies. The monoclonal antibodies were characterized for binding affinity, epitope mapping by pairing analysis, and nucleotide and amino acid sequences. AChE was solubilized from frozen RBCs with 1% (v/v) Triton X-100. A 16 mL sample containing 5.8 μg of RBC AChE was treated with a quantity of soman model compound that inhibited 50% of the AChE activity. Native and soman-inhibited RBC AChE samples were immunopurified on antibody-Sepharose beads. The immunopurified RBC AChE was digested with pepsin and analyzed by liquid chromatography tandem mass spectrometry on a 6600 Triple-TOF mass spectrometer. The aged soman-modified PheGlyGluSerAlaGlyAlaAlaSer (FGESAGAAS) peptide was detected using a targeted analysis method. It was concluded that all 6 monoclonal antibodies could be used to immunopurify RBC AChE and that exposure to nerve agents could be detected as adducts on the active site serine of RBC AChE.
Sigma-Aldrich
Anti-Acetylcholinesterase Antibody, clone AE-1, clone AE-1, Chemicon®, from mouse