Merck
  • Thrombin induces tumor invasion through the induction and association of matrix metalloproteinase-9 and beta1-integrin on the cell surface.
Thrombin induces tumor invasion through the induction and association of matrix metalloproteinase-9 and beta1-integrin on the cell surface.
The Journal of biological chemistry (2007-12-01)
A Reza Radjabi, Kenjiro Sawada, Sujatha Jagadeeswaran, Alfred Eichbichler, Hilary A Kenny, Anthony Montag, Katharina Bruno, Ernst Lengyel
ABSTRACT
The procoagulatory serine protease, thrombin, is known to induce invasion and metastasis in various cancers, but the mechanisms by which it promotes tumorigenesis are poorly understood. Because the 92-kDa gelatinase (MMP-9) is a known mediator of tumor cell invasion, we sought to determine whether and how thrombin regulates MMP-9. The thrombin receptor, PAR-1, and MMP-9 are expressed in osteosarcomas, as determined by immunohistochemistry. Stimulation of U2-OS osteosarcoma cells with thrombin and a thrombin receptor-activating peptide induced pro-MMP-9 secretion as well as cell surface-associated pro-MMP-9 expression and proteolytic activity. This was paralleled by an increase in MMP-9 mRNA and MMP-9 promoter activity. Thrombin-induced invasion of U2-OS cells through Matrigel was mediated by the phosphatidylinositol 3-kinase signaling pathway and could be inhibited with an MMP-9 antibody. The stimulation of MMP-9 by thrombin was paralleled by an increase in beta1-integrin mRNA and beta1-integrin expression on the cell surface, which was also mediated by phosphatidylinositol 3-kinase and was required for invasion. Thrombin activation induced and co-localized both beta1-integrin and pro-MMP-9 on the cell membrane, as evidenced by co-immunoprecipitation, confocal microscopy, and a protein binding assay. The thrombin-mediated association of these two proteins, as well as thrombin-mediated invasion of U2-OS cells, could be blocked with a cyclic peptide and with an antibody preventing binding of the MMP-9 hemopexin domain to beta1-integrin. These results suggest that thrombin induces expression and association of beta1-integrin with MMP-9 and that the cell surface localization of the protease by the integrin promotes tumor cell invasion.
Sigma-Aldrich
Anti-Integrin α1 Antibody, clone FB12, clone FB12, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin β3 Antibody, clone B3A, azide free, clone B3A, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin αVβ5 Antibody, clone P1F6, clone P1F6, Chemicon®, from mouse