Merck
  • Beta ig-h3 promotes renal proximal tubular epithelial cell adhesion, migration and proliferation through the interaction with alpha3beta1 integrin.
Beta ig-h3 promotes renal proximal tubular epithelial cell adhesion, migration and proliferation through the interaction with alpha3beta1 integrin.
Experimental & molecular medicine (2004-07-24)
Sun-Woo Park, Jong-Sup Bae, Ki-San Kim, Sun-Hee Park, Byung-Heon Lee, Je-Yong Choi, Jae-Yong Park, Sung-Woo Ha, Yong-Lim Kim, Tae-Hwan Kwon, In-San Kim, Rang-Woon Park
ABSTRACT
Betaig-h3 (betaig-h3) is a secretory protein composed of fasciclin I-like repeats containing sequences that allows binding of integrins and glycosaminoglycans in vivo. Expression of betaig-h3 is responsive to TGF-Beta and the protein is found to be associated with extracellular matrix (ECM) molecules, implicating betaig-h3 as an ECM adhesive protein of developmental processes. We previously observed predominant expression of betaig-h3 expression in the basement membrane of proximal tubules of kidney. In this study, the physiological relevance of such localized expression of betaig-h3 was examined in the renal proximal tubular epithelial cells (RPTEC). RPTEC constitutively expressed betaig-h3 and the expression was dramatically induced by exogenous TGF-Beta1 treatment. betaig-h3 and its second and fourth FAS1 domain were able to mediate RPTEC adhesion, spreading and migration. Two known alpha3beta1 integrin-interaction motifs including aspartatic acid and isoleucine residues, NKDIL and EPDIM in betaig-h3 were responsible to mediate RPTEC adhesion, spreading, and migration. By using specific antibodies against integrins, we confirmed that alpha3beta1 integrin mediates the adhesion and migration of RPTECs on betaig-h3. In addition, it also enhanced proliferation of RPTECs through NKDIL and EPDIM. These results indicate that betaig-h3 mediates adhesion, spreading, migration and proliferation of RPTECs through the interaction with alpha3beta1 integrin and is intimately involved in the maintenance and the regeneration of renal proximal tubular epithelium.
Sigma-Aldrich
Anti-Integrin αVβ3 Antibody, clone LM609, clone LM609, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin α4 Antibody, clone P1H4, clone P1H4, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin αVβ5 Antibody, clone P1F6, clone P1F6, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin α1 Antibody, clone FB12, clone FB12, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin β2 Antibody, clone P4H9, clone P4H9, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin α2 Antibody, clone P1E6, clone P1E6, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin β1 Antibody, clone 6S6, clone 6S6, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin α6 Antibody, clone NKI-GoH3, clone NKI-GoH3, Chemicon®, from rat
Sigma-Aldrich
Anti-Integrin αV Antibody, clone P3G8, clone P3G8, Chemicon®, from mouse