Merck
JAM2 interacts with alpha4beta1. Facilitation by JAM3.
The Journal of biological chemistry (2002-06-19)
Sonia A Cunningham, Jose M Rodriguez, M Pia Arrate, Tuan M Tran, Tommy A Brock
ABSTRACT
We have previously reported that junctional adhesion molecule 2 (JAM2) adheres to T cells through heterotypic interactions with JAM3. An examination of the cation dependence of JAM2 adhesion to HSB cells revealed a Mn(2+)-enhanced binding component indicative of integrin involvement. Using neutralizing integrin antibodies, we have defined an interaction between JAM2 and alpha(4)beta(1) in T cells. The interaction is readily amenable to drug intervention as demonstrated by the ability of TBC 772, an alpha(4)-specific inhibitor, to attenuate the Mn(2+)-enhanced component. Intriguingly, the engagement of alpha(4)beta(1) by JAM2 is only enabled following prior adhesion of JAM2 with JAM3 and is not detectable in cells where JAM3 expression is absent. Supporting this observation, we show that neutralizing JAM3 serum and soluble JAM3 ectodomain inhibit not only JAM2 binding to JAM3 but also prevent JAM2/alpha(4)beta(1) interactions in T cells. We further define the first Ig-like fold of JAM2 as being competent in binding both JAM3 and alpha(4)beta(1) counter-receptors. Mutagenesis of the only acidic residue in the C-D loop of this Ig fold, namely Asp-82, has no bearing on alpha(4)beta(1) interactions, and thus JAM2 deviates somewhat from the mechanism used by other immunoglobulin superfamily cell adhesion molecules to engage integrin.
Sigma-Aldrich
Anti-Integrin β1 Antibody, clone P5D2, clone P5D2, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin α4 Antibody, clone P4C2, ascites fluid, clone P4C2, Chemicon®