Merck
  • Experimental evidence for the age dependence of tau protein spread in the brain.
Experimental evidence for the age dependence of tau protein spread in the brain.
Science advances (2019-06-30)
Susanne Wegmann, Rachel E Bennett, Louis Delorme, Ashley B Robbins, Miwei Hu, Danny McKenzie, Molly J Kirk, Julia Schiantarelli, Nahel Tunio, Ana C Amaral, Zhanyun Fan, Samantha Nicholls, Eloise Hudry, Bradley T Hyman
ABSTRACT
The incidence of Alzheimer's disease (AD), which is characterized by progressive cognitive decline that correlates with the spread of tau protein aggregation in the cortical mantle, is strongly age-related. It could be that age predisposes the brain for tau misfolding and supports the propagation of tau pathology. We tested this hypothesis using an experimental setup that allowed for exploration of age-related factors of tau spread and regional vulnerability. We virally expressed human tau locally in entorhinal cortex (EC) neurons of young or old mice and monitored the cell-to-cell tau protein spread by immunolabeling. Old animals showed more tau spreading in the hippocampus and adjacent cortical areas and accumulated more misfolded tau in EC neurons. No misfolding, at any age, was observed in the striatum, a brain region mostly unaffected by tangles. Age and brain region dependent tau spreading and misfolding likely contribute to the profound age-related risk for sporadic AD.
Sigma-Aldrich
Anti-Actin Antibody, clone C4, clone C4, Chemicon®, from mouse
Sigma-Aldrich
Anti-Synapsin-1 Antibody, clone 10.22, clone 10.22, from mouse