Merck
  • Alfacalcidol treatment restores derailed immune-regulation in patients with undifferentiated connective tissue disease.
Alfacalcidol treatment restores derailed immune-regulation in patients with undifferentiated connective tissue disease.
Autoimmunity reviews (2010-09-28)
Eva Zold, Peter Szodoray, Britt Nakken, Sandor Barath, Janos Kappelmayer, Laszlo Csathy, Agota Hajas, Sandor Sipka, Edit Gyimesi, Janos Gaal, Zsolt Barta, Judit Hallay, Gyula Szegedi, Edit Bodolay
ABSTRACT
Vitamin D deficiency may contribute to pathological changes in the number and function of CD4+ T helper cell subsets (CD4+Th1, CD4+Th17, CD4+CD25(bright)Foxp3-natural regulatory T cells-nTreg) in patients with undifferentiated connective tissue disease (UCTD). The aim of the present study was to evaluate, whether alfacalcidol could restore immune-regulatory changes in patients with UCTD. We assessed the optimal dose of alfacalcidol that could normalize the elevated levels of IFN-γ expressed by the CD4+Th1 cells and the IL-17 expressed by Th17 cells. Furthermore alfacalcidol decreased the Th1 and Th17 related cytokine levels, repaired the nTreg/Th7 balance, and restored the functional activity of nTreg cells. Twenty one UCTD patients with Vitamin D deficiency (<30 ng/ml) were administered with three different daily doses of alfacalcidol. Seven patients were supplemented with 0.5 μg/day, 7 patients with 1.0 μg/day, and 7 patients with 1.5 μg/day alfacalcidol treatment during 5 weeks. Our results indicated that 1.0 μg/day alfacalcidol during 5 weeks was the optimal therapeutic regime to increase the vitamin D levels, repair the nTreg/Th17 balance and raise the capacity of nTreg cells to suppress the proliferation of autologous CD4+CD25- cells. 1.5 μg daily dose alfacalcidol was not more effective than the 1.0 μg/day treatment. In this study we described that vitamin D deficiency can contribute to the complex immune-regulatory abnormalities in patients with UCTD and vitamin D substitution therapy can improve the fine balance of pro- and anti-inflammatory processes in the disease.
Sigma-Aldrich
1α-Hydroxyvitamin D3, ≥97.0% (HPLC)