Merck
  • Wnt7b signalling through Frizzled-7 receptor promotes dendrite development by coactivating CaMKII and JNK.
Wnt7b signalling through Frizzled-7 receptor promotes dendrite development by coactivating CaMKII and JNK.
Journal of cell science (2018-06-15)
María E Ferrari, María E Bernis, Faye McLeod, Marina Podpolny, Romina P Coullery, Inelia M Casadei, Patricia C Salinas, Silvana B Rosso
ABSTRACT
The formation of complex dendritic arbors is crucial for the assembly of functional networks as abnormal dendrite formation underlies several neurodevelopmental and psychiatric disorders. Many extracellular factors have been postulated as regulators of dendritic growth. Wnt proteins play a critical role in neuronal development and circuit formation. We previously demonstrated that Wnt7b acts through the scaffold protein dishevelled 1 (Dvl1) to modulate dendrite arborisation by activating a non-canonical Wnt signalling pathway. Here, we identify the seven-transmembrane frizzled-7 (Fz7, also known as FZD7) as the receptor for Wnt7b-mediated dendrite growth and complexity. Importantly, Fz7 is developmentally regulated in the intact hippocampus, and is localised along neurites and at dendritic growth cones, suggesting a role in dendrite formation and maturation. Fz7 loss-of-function studies demonstrated that Wnt7b requires Fz7 to promote dendritic arborisation. Moreover, in vivo Fz7 loss of function results in dendritic defects in the intact mouse hippocampus. Furthermore, our findings reveal that Wnt7b and Fz7 induce the phosphorylation of Ca2+/calmodulin-dependent protein kinase II (CaMKII) and JNK proteins, which are required for dendritic development. Here, we demonstrate that Wnt7b-Fz7 signals through two non-canonical Wnt pathways to modulate dendritic growth and complexity.
Sigma-Aldrich
Anti-β-Tubulin III antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Frizzled-7 Antibody, from rabbit, purified by affinity chromatography
Pricing and availability is not currently available.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-MAP2 (2a+2b) antibody, Mouse monoclonal, clone AP-20, ascites fluid
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-c-Myc antibody, Mouse monoclonal, clone 9E10, purified from hybridoma cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
MONOCLONAL ANTI-MYC TAG antibody produced in mouse, clone 9E10, IgG fraction of antiserum, buffered aqueous solution
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Tau-1 Antibody, clone PC1C6, clone PC1C6, Chemicon®, from mouse
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-GFP (Green Fluorescent Protein) Antibody, Upstate®, from chicken
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-MYC (Ab-58) antibody produced in rabbit, affinity isolated antibody
Pricing and availability is not currently available.