Merck
  • Brain tyrosinase overexpression implicates age-dependent neuromelanin production in Parkinson's disease pathogenesis.
Brain tyrosinase overexpression implicates age-dependent neuromelanin production in Parkinson's disease pathogenesis.
Nature communications (2019-03-09)
Iria Carballo-Carbajal, Ariadna Laguna, Jordi Romero-Giménez, Thais Cuadros, Jordi Bové, Marta Martinez-Vicente, Annabelle Parent, Marta Gonzalez-Sepulveda, Núria Peñuelas, Albert Torra, Beatriz Rodríguez-Galván, Andrea Ballabio, Takafumi Hasegawa, Analía Bortolozzi, Ellen Gelpi, Miquel Vila
ABSTRACT
In Parkinson's disease (PD) there is a selective degeneration of neuromelanin-containing neurons, especially substantia nigra dopaminergic neurons. In humans, neuromelanin accumulates with age, the latter being the main risk factor for PD. The contribution of neuromelanin to PD pathogenesis remains unknown because, unlike humans, common laboratory animals lack neuromelanin. Synthesis of peripheral melanins is mediated by tyrosinase, an enzyme also present at low levels in the brain. Here we report that overexpression of human tyrosinase in rat substantia nigra results in age-dependent production of human-like neuromelanin within nigral dopaminergic neurons, up to levels reached in elderly humans. In these animals, intracellular neuromelanin accumulation above a specific threshold is associated to an age-dependent PD phenotype, including hypokinesia, Lewy body-like formation and nigrostriatal neurodegeneration. Enhancing lysosomal proteostasis reduces intracellular neuromelanin and prevents neurodegeneration in tyrosinase-overexpressing animals. Our results suggest that intracellular neuromelanin levels may set the threshold for the initiation of PD.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Monoclonal Anti-Glial Fibrillary Acidic Protein (GFAP) antibody produced in mouse, clone G-A-5, ascites fluid
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Duolink® In Situ Probemaker MINUS
Sigma-Aldrich
Duolink® In Situ Detection Reagents Red
Sigma-Aldrich
Duolink® In Situ Probemaker PLUS
Sigma-Aldrich
Anti-Tyrosine Hydroxylase Rabbit pAb, liquid, Calbiochem®