Merck
  • Protective outcomes of low-dose doxycycline on renal function of Wistar rats subjected to acute ischemia/reperfusion injury.
Protective outcomes of low-dose doxycycline on renal function of Wistar rats subjected to acute ischemia/reperfusion injury.
Biochimica et biophysica acta. Molecular basis of disease (2017-10-11)
Aline L Cortes, Sabrina R Gonsalez, Lilimar S Rioja, Simone S C Oliveira, André L S Santos, Minolfa C Prieto, Paulo A Melo, Lucienne S Lara
ABSTRACT
Renal ischemia-reperfusion injury (IRI) is a major cause of acute renal failure. Doxycycline (Dc) belongs to the tetracycline-class of antibiotics with demonstrated beneficial molecular effects in the brain and heart, mainly through matrix metalloproteinases inhibition (MMP). However, Dc protection of renal function has not been demonstrated. We determined whether low doses of Dc would prevent decreases in glomerular filtration rate (GFR) and maintain tubular Na+ handling in Wistar rats subjected to kidney I/R. Male Wistar rats underwent bilateral kidney ischemia for 30min followed by 24h reperfusion (I/R). Doxycycline (1, 3, and 10mg/kg, i.p.) was administered 2h before surgery. Untreated I/R rats showed a 250% increase in urine volume and proteinuria, a 60% reduction in GFR, accumulation of urea-nitrogen in the blood, and a 60% decrease in the fractional Na+ excretion due to unbalanced Na+ transporter activity. Treatment with Dc 3mg/kg maintained control levels of urine volume, proteinuria, GFR, blood urea-nitrogen, fractional Na+ excretion, and equilibrated Na+ transporter activities. The Dc protection effects on renal function were associated with kidney structure preservation and prevention of TGFβ and fibronectin deposition. In vitro, total MMP activity was augmented in I/R and inhibited by 25 and 50μM Dc. In vivo, I/R augmented MMP-2 and -9 protein content without changing their activities. Doxycycline treatment downregulated total MMP activity and MMP-2 and -9 protein content. Our results suggest that treatment with low dose Dc protects from IRI, thereby preserving kidney function.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Sigma-Aldrich
Z-Arg-Arg-7-amido-4-methylcoumarin hydrochloride