Merck
  • TRIM5α Restricts Flavivirus Replication by Targeting the Viral Protease for Proteasomal Degradation.
TRIM5α Restricts Flavivirus Replication by Targeting the Viral Protease for Proteasomal Degradation.
Cell reports (2019-06-13)
Abhilash I Chiramel, Nicholas R Meyerson, Kristin L McNally, Rebecca M Broeckel, Vanessa R Montoya, Omayra Méndez-Solís, Shelly J Robertson, Gail L Sturdevant, Kirk J Lubick, Vinod Nair, Brian H Youseff, Robin M Ireland, Catharine M Bosio, Kyusik Kim, Jeremy Luban, Vanessa M Hirsch, R Travis Taylor, Fadila Bouamr, Sara L Sawyer, Sonja M Best
ABSTRACT
Tripartite motif-containing protein 5α (TRIM5α) is a cellular antiviral restriction factor that prevents early events in retrovirus replication. The activity of TRIM5α is thought to be limited to retroviruses as a result of highly specific interactions with capsid lattices. In contrast to this current understanding, we show that both human and rhesus macaque TRIM5α suppress replication of specific flaviviruses. Multiple viruses in the tick-borne encephalitis complex are sensitive to TRIM5α-dependent restriction, but mosquito-borne flaviviruses, including yellow fever, dengue, and Zika viruses, are resistant. TRIM5α suppresses replication by binding to the viral protease NS2B/3 to promote its K48-linked ubiquitination and proteasomal degradation. Importantly, TRIM5α contributes to the antiviral function of IFN-I against sensitive flaviviruses in human cells. Thus, TRIM5α possesses remarkable plasticity in the recognition of diverse virus families, with the potential to influence human susceptibility to emerging flaviviruses of global concern.
Sigma-Aldrich
Epoxomicin, ≥95% (HPLC), solid
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Bafilomycin A1 from Streptomyces griseus, ≥90% (HPLC)
Sigma-Aldrich
Rabbit Anti-Chicken IgY Antibody, HRP conjugate, Upstate®, from rabbit
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial
Roche
Anti-HA-Peroxidase, High Affinity, from rat IgG1
Sigma-Aldrich
Puromycin dihydrochloride from Streptomyces alboniger, powder, BioReagent, suitable for cell culture