Merck
Penicillin-induced lysis of Streptococcus mutans.
Infection and immunity (1984-11-01)
T A Kral, M D Callaway
ABSTRACT
Treatment of Streptococcus mutans GS-5 cells with concentrations of penicillin G within a relatively narrow range resulted in substantial lysis. This penicillin-induced lysis was dependent upon cell density and pH of the lysis medium. Other oral streptococci (Streptococcus sobrinus, Streptococcus rattus, and Streptococcus cricetus) also demonstrated substantial levels of penicillin-induced lysis under appropriate conditions. Lesser degrees of lysis were seen in a related organism, Streptococcus ferus.
Sigma-Aldrich
Penicillin G potassium salt, 95.0 - 102.0%
Pricing and availability is not currently available.