Merck
  • The SS18-SSX Oncoprotein Hijacks KDM2B-PRC1.1 to Drive Synovial Sarcoma.
The SS18-SSX Oncoprotein Hijacks KDM2B-PRC1.1 to Drive Synovial Sarcoma.
Cancer cell (2018-03-06)
Ana Banito, Xiang Li, Aimée N Laporte, Jae-Seok Roe, Francisco Sanchez-Vega, Chun-Hao Huang, Amanda R Dancsok, Katerina Hatzi, Chi-Chao Chen, Darjus F Tschaharganeh, Rohit Chandwani, Nilgun Tasdemir, Kevin B Jones, Mario R Capecchi, Christopher R Vakoc, Nikolaus Schultz, Marc Ladanyi, Torsten O Nielsen, Scott W Lowe
ABSTRACT
Synovial sarcoma is an aggressive cancer invariably associated with a chromosomal translocation involving genes encoding the SWI-SNF complex component SS18 and an SSX (SSX1 or SSX2) transcriptional repressor. Using functional genomics, we identify KDM2B, a histone demethylase and component of a non-canonical polycomb repressive complex 1 (PRC1.1), as selectively required for sustaining synovial sarcoma cell transformation. SS18-SSX1 physically interacts with PRC1.1 and co-associates with SWI/SNF and KDM2B complexes on unmethylated CpG islands. Via KDM2B, SS18-SSX1 binds and aberrantly activates expression of developmentally regulated genes otherwise targets of polycomb-mediated repression, which is restored upon KDM2B depletion, leading to irreversible mesenchymal differentiation. Thus, SS18-SSX1 deregulates developmental programs to drive transformation by hijacking a transcriptional repressive complex to aberrantly activate gene expression.
Sigma-Aldrich
Anti-trimethyl-Histone H3 (Lys27) Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-JHDM1B Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-Bmi-1 Antibody, clone F6, clone F6, Upstate®, from mouse
Sigma-Aldrich
ChIPAb+ JHDM1B Antibody, from rabbit, purified by affinity chromatography