Merck
  • The binding of captopril to angiotensin I-converting enzyme triggers activation of signaling pathways.
The binding of captopril to angiotensin I-converting enzyme triggers activation of signaling pathways.
American journal of physiology. Cell physiology (2018-06-07)
Rosana I Reis, Marie D Nogueira, Ana Lucia Campanha-Rodrigues, Larissa Miranda Pereira, Maria Claudina C Andrade, Lucas T Parreiras-E-Silva, Claudio M Costa-Neto, Renato Arruda Mortara, Dulce E Casarini
ABSTRACT
Hypertension is a global health problem, and angiotensin I (ANG I)-converting enzyme (ACE) inhibitors are largely used to control this pathology. Recently, it has been shown that ACE can also act as a transducer signal molecule when its inhibitors or substrates bind to it. This new role of ACE could contribute to understanding some of the effects not explained by its catalytic activity only. In this study, we investigated signaling pathway activation in Chinese hamster ovary (CHO) cells stably expressing ACE (CHO-ACE) under different conditions. We also investigated gene modulation after 4 h and 24 h of captopril treatment. Our results demonstrated that CHO-ACE cells when stimulated with ANG I, ramipril, or captopril led to JNK and ERK1/2 phosphorylation. To verify any physiological role at the endogenous level, we made use of primary cultures of mesangial cells from spontaneously hypertensive rats (SHR) and Wistar rats. Our results showed that ERK1/2 activation occurred mainly in primary cultures of mesangial cells from SHR rats upon captopril stimulation, suggesting that this signaling pathway could be differentially regulated during hypertension. Our results also showed that captopril treatment leads to a decrease of cyclooxygenase 2, interleukin-1β, and β-arrestin2 and a significant increase of AP2 gene expression levels. Our findings strengthen the fact that, in addition to the blockage of enzymatic activity, ACE inhibitors also trigger signaling pathway activation, and this may contribute to their beneficial effects in the treatment of hypertension and other pathologies.
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody
Sigma-Aldrich
Anti-Angiotensin Converting Enzyme Antibody, clone 9B9, clone 9B9, Chemicon®, from mouse
Sigma-Aldrich
SP600125, ≥98% (HPLC)
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution