Merck
  • Inhibition of Discoidin Domain Receptor 1 Reduces Collagen-mediated Tumorigenicity in Pancreatic Ductal Adenocarcinoma.
Inhibition of Discoidin Domain Receptor 1 Reduces Collagen-mediated Tumorigenicity in Pancreatic Ductal Adenocarcinoma.
Molecular cancer therapeutics (2017-09-03)
Kristina Y Aguilera, Huocong Huang, Wenting Du, Moriah M Hagopian, Zhen Wang, Stefan Hinz, Tae Hyun Hwang, Huamin Wang, Jason B Fleming, Diego H Castrillon, Xiaomei Ren, Ke Ding, Rolf A Brekken
ABSTRACT
The extracellular matrix (ECM), a principal component of pancreatic ductal adenocarcinoma (PDA), is rich in fibrillar collagens that facilitate tumor cell survival and chemoresistance. Discoidin domain receptor 1 (DDR1) is a receptor tyrosine kinase that specifically binds fibrillar collagens and has been implicated in promoting cell proliferation, migration, adhesion, ECM remodeling, and response to growth factors. We found that collagen-induced activation of DDR1 stimulated protumorigenic signaling through protein tyrosine kinase 2 (PYK2) and pseudopodium-enriched atypical kinase 1 (PEAK1) in pancreatic cancer cells. Pharmacologic inhibition of DDR1 with an ATP-competitive orally available small-molecule kinase inhibitor (7rh) abrogated collagen-induced DDR1 signaling in pancreatic tumor cells and consequently reduced colony formation and migration. Furthermore, the inhibition of DDR1 with 7rh showed striking efficacy in combination with chemotherapy in orthotopic xenografts and autochthonous pancreatic tumors where it significantly reduced DDR1 activation and downstream signaling, reduced primary tumor burden, and improved chemoresponse. These data demonstrate that targeting collagen signaling in conjunction with conventional cytotoxic chemotherapy has the potential to improve outcome for pancreatic cancer patients. Mol Cancer Ther; 16(11); 2473-85. ©2017 AACR.
Sigma-Aldrich
Anti-PEAK1 Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-phospho-Peak Antibody (Tyr665), from rabbit, purified by affinity chromatography
Sigma-Aldrich
DDR1 Inhibitor 7rh, ≥98% (HPLC)