Merck
  • Association of insulin receptor substrate proteins with Bcl-2 and their effects on its phosphorylation and antiapoptotic function.
Association of insulin receptor substrate proteins with Bcl-2 and their effects on its phosphorylation and antiapoptotic function.
Molecular biology of the cell (2000-02-26)
H Ueno, E Kondo, R Yamamoto-Honda, K Tobe, T Nakamoto, K Sasaki, K Mitani, A Furusaka, T Tanaka, Y Tsujimoto, T Kadowaki, H Hirai
ABSTRACT
Insulin receptor substrate (IRS) proteins are docking proteins that couple growth factor receptors to various effector molecules, including phosphoinositide-3 kinase, Grb-2, Syp, and Nck. Here we show that IRS-1 associates with the loop domain of Bcl-2 and synergistically up-regulates antiapoptotic function of Bcl-2. IRS-2 but not IRS-3 binds to Bcl-2, and IRS-1 associates with Bcl-XL but not with Bax or Bik. Overexpression of IRS-1 suppresses phosphorylation of Bcl-2 induced by stimulation with insulin, and the hypophosphorylation may lead to its enhanced antiapoptotic activity. The binding site for Bcl-2 is located on the carboxyl half-domain of IRS-1. IRS-3, which lacks the corresponding region, dominant-negatively abrogates the survival effects of IRS-1 and Bcl-2. For the antiapoptotic activity of IRS-1, binding to Bcl-2 is more critical than activating phosphoinositide-3 kinase. Our results indicate that IRS proteins transmit signals from the insulin receptor to Bcl-2, thus regulating cell survival probably through regulating phosphorylation of Bcl-2.
Sigma-Aldrich
Anti-Phosphotyrosine Antibody, clone 4G10®, clone 4G10®, Upstate®, from mouse