Merck
  • Steady-state increase of cAMP-response element binding protein, Rac, and PAK signaling in presenilin-deficient neurons.
Steady-state increase of cAMP-response element binding protein, Rac, and PAK signaling in presenilin-deficient neurons.
Journal of neurochemistry (2007-11-13)
Natalie Y Barnes, Jun Shi, Hiroshi Yajima, Gopal Thinakaran, Angèle T Parent
ABSTRACT
Mutations in the genes encoding presenilins (PS1 and PS2) account for the majority of cases of early-onset Alzheimer's disease. PS1 and PS2 form the catalytic center of gamma-secretase, an enzyme responsible for intramembraneous proteolysis of several type I transmembrane proteins. Many gamma-secretase substrates are coupled to intracellular signaling events such as cAMP-response element binding protein and Rac1/p21-activated kinase pathways, which are associated with synaptic function. Here, we have examined the activation of these pathways in neurons lacking PS1 expression or gamma-secretase activity. We found evidence for heightened steady-state activation of cAMP-response element binding protein, Rac1, and p21-activated kinase signaling in PS-deficient neurons. Our study highlights the importance of PS-dependent proteolytic cleavage of gamma-secretase substrates in regulating neuronal signal transduction.
Sigma-Aldrich
Anti-PSD95 Antibody, clone K28/43, clone K28/43, from mouse