Merck
  • Identification of Eph receptor signaling as a regulator of autophagy and a therapeutic target in colorectal carcinoma.
Identification of Eph receptor signaling as a regulator of autophagy and a therapeutic target in colorectal carcinoma.
Molecular oncology (2019-09-24)
Michael DiPrima, Dunrui Wang, Alix Tröster, Dragan Maric, Nekane Terrades-Garcia, Taekyu Ha, Hyeongil Kwak, David Sanchez-Martin, Denis Kudlinzki, Harald Schwalbe, Giovanna Tosato
ABSTRACT
Advanced colorectal carcinoma is currently incurable, and new therapies are urgently needed. We report that phosphotyrosine-dependent Eph receptor signaling sustains colorectal carcinoma cell survival, thereby uncovering a survival pathway active in colorectal carcinoma cells. We find that genetic and biochemical inhibition of Eph tyrosine kinase activity or depletion of the Eph ligand EphrinB2 reproducibly induces colorectal carcinoma cell death by autophagy. Spautin and 3-methyladenine, inhibitors of early steps in the autophagic pathway, significantly reduce autophagy-mediated cell death that follows inhibition of phosphotyrosine-dependent Eph signaling in colorectal cancer cells. A small-molecule inhibitor of the Eph kinase, NVP-BHG712 or its regioisomer NVP-Iso, reduces human colorectal cancer cell growth in vitro and tumor growth in mice. Colorectal cancers express the EphrinB ligand and its Eph receptors at significantly higher levels than numerous other cancer types, supporting Eph signaling inhibition as a potential new strategy for the broad treatment of colorectal carcinoma.
Sigma-Aldrich
Rapamycin, Ready Made Solution, 2.5 mg/mL in DMSO (2.74 mM), from Streptomyces hygroscopicus
Sigma-Aldrich
Spautin-1, ≥98% (HPLC)
Sigma-Aldrich
O-Phospho-L-tyrosine
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Chloroquine diphosphate salt, powder or crystals, 98.5-101.0% (EP)
Sigma-Aldrich
(+)-Sodium L-ascorbate, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
TWEEN® 20, viscous liquid
Sigma-Aldrich
Sodium orthovanadate, ≥90% (titration)
Sigma-Aldrich
3-Methyladenine, autophagy inhibitor
Sigma-Aldrich
2-Mercaptoethanol, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 2, aqueous solution (dark coloration may develop upon storage, which does not affect the activity)
Sigma-Aldrich
NVP-BHG712, ≥98% (HPLC)
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Sigma-Aldrich
Newborn Calf Serum, USA origin, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Heparin sodium salt from porcine intestinal mucosa, Grade I-A, ≥180 USP units/mg, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Triton X-100, BioXtra
Sigma-Aldrich
ECO TWEEN® 20, viscous liquid
Sigma-Aldrich
Donkey serum