Merck
  • Both maltose-binding protein and ATP are required for nucleotide-binding domain closure in the intact maltose ABC transporter.
Both maltose-binding protein and ATP are required for nucleotide-binding domain closure in the intact maltose ABC transporter.
Proceedings of the National Academy of Sciences of the United States of America (2008-08-30)
Cedric Orelle, Tulin Ayvaz, R Michael Everly, Candice S Klug, Amy L Davidson
ABSTRACT
The maltose transporter MalFGK(2) of Escherichia coli is a member of the ATP-binding cassette superfamily. A periplasmic maltose-binding protein (MBP) delivers maltose to MalFGK(2) and stimulates its ATPase activity. Site-directed spin labeling EPR spectroscopy was used to study the opening and closing of the nucleotide-binding interface of MalFGK(2) during the catalytic cycle. In the intact transporter, closure of the interface coincides not just with the binding of ATP, as seen with isolated nucleotide-binding domains, but requires both MBP and ATP, implying that MBP stimulates ATPase activity by promoting the closure of the nucleotide-binding interface. After ATP hydrolysis, with MgADP and MBP bound, the nucleotide-binding interface resides in a semi-open configuration distinct from the fully open configuration seen in the absence of any ligand. We propose that P(i) release coincides with the reorientation of transmembrane helices to an inward-facing conformation and the final step of maltose translocation into the cell.
Sigma-Aldrich
L-α-Phosphatidylcholine, from soybean, Type II-S, 14-29% choline basis