Merck
  • Stereoelectronic Effects in Ligand Design: Enantioselective Rhodium-Catalyzed Hydrogenation of Aliphatic Cyclic Tetrasubstituted Enamides and Concise Synthesis of (R)-Tofacitinib.
Stereoelectronic Effects in Ligand Design: Enantioselective Rhodium-Catalyzed Hydrogenation of Aliphatic Cyclic Tetrasubstituted Enamides and Concise Synthesis of (R)-Tofacitinib.
Angewandte Chemie (International ed. in English) (2019-07-26)
Chengxi Li, Feng Wan, Yuan Chen, Henian Peng, Wenjun Tang, Shu Yu, J Christopher McWilliams, Jason Mustakis, Lacey Samp, Robert J Maguire
ABSTRACT
We herein report the development of a conformationally defined, electron-rich, C2 -symmetric, P-chiral bisphosphorus ligand, ArcPhos, by taking advantage of stereoelectronic effects in ligand design. With the Rh-ArcPhos catalyst, excellent enantioselectivities and unprecedentedly high turnovers (TON up to 10 000) were achieved in the asymmetric hydrogenation of aliphatic carbocyclic and heterocyclic tetrasubstituted enamides, to generate a series of chiral cis-2-alkyl-substituted carbocyclic and heterocyclic amine derivatives in excellent enantiomeric ratios. This method also enabled an efficient and practical synthesis of the Janus kinase inhibitor (R)-tofacitinib.
Sigma-Aldrich
(2R,2′R,3R,3′R)-MeO-BIBOP
Pricing and availability is not currently available.