Merck
  • Probiotic Supplementation is Associated with Increased Antioxidant Capacity and Copper Chelation in C. difficile-Infected Fecal Water.
Probiotic Supplementation is Associated with Increased Antioxidant Capacity and Copper Chelation in C. difficile-Infected Fecal Water.
Nutrients (2019-08-29)
Mohd Baasir Gaisawat, Michèle M Iskandar, Chad W MacPherson, Thomas A Tompkins, Stan Kubow
ABSTRACT
Probiotic supplementation plays a key role in maintaining intestinal homeostasis due to its ability to modulate gut microbiota. Although their potential as potent antioxidants have previously been explored, their ability to affect the redox status in the gut lumen of healthy subjects or those with gastrointestinal (GI) disorders remains unclear. In our study, we assessed the ability of single strain and multispecies probiotic supplementation to cause a change in the redox status of normal fecal water and in Clostridium (C.) difficile-infected fecal water using a simulated gastrointestinal model. Changes in redox status were assessed by ferric-reducing antioxidant power (FRAP), 2',2'-diphenyl-1-picrylhydrazyl (DPPH), and iron and copper chelation assays. The findings from our study showed that in normal fecal water, probiotic supplements, apart from Lactobacillus (L.) rhamnosus R0011, showed a significant increase in iron chelation (p < 0.05), which was associated with lower FRAP and copper chelation. In C. difficile-infected fecal water, all probiotic supplements showed a significant increase in FRAP (p < 0.05) and were associated with increased copper chelation. The DPPH assay showed no treatment effect in either fecal water. These findings suggest that C. difficile mediates dysregulation of redox status, which is counteracted by probiotics through ferric-reducing ability and copper chelation.
Sigma-Aldrich
Sodium acetate trihydrate, BioXtra, ≥99.0%
Sigma-Aldrich
DL-Cysteine, technical grade
Sigma-Aldrich
L-Cysteine, 97%
Sigma-Aldrich
Pyrocatechol Violet, suitable for indicator
Sigma-Aldrich
N-(1-Naphthyl)ethylenediamine dihydrochloride, ≥98%
Supelco
2,4,6-Tris(2-pyridyl)-s-triazine, ≥98%
Sigma-Aldrich
Salicylic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium nitrate, BioXtra, ≥99.0%
Sigma-Aldrich
Sulfanilamide, ≥98%
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, reagent grade, 98.5-101.5% (titration)
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
Iron(III) chloride hexahydrate, ACS reagent, 97%