Merck
  • Ikaros prevents autoimmunity by controlling anergy and Toll-like receptor signaling in B cells.
Ikaros prevents autoimmunity by controlling anergy and Toll-like receptor signaling in B cells.
Nature immunology (2019-10-09)
Tanja A Schwickert, Hiromi Tagoh, Karina Schindler, Maria Fischer, Markus Jaritz, Meinrad Busslinger
ABSTRACT
The establishment of a diverse B cell antigen receptor (BCR) repertoire by V(D)J recombination also generates autoreactive B cells. Anergy is one tolerance mechanism; it renders autoreactive B cells insensitive to stimulation by self-antigen, whereas Toll-like receptor (TLR) signaling can reactivate anergic B cells. Here, we describe a critical role of the transcription factor Ikaros in controlling BCR anergy and TLR signaling. Mice with specific deletion of Ikaros in mature B cells developed systemic autoimmunity. Ikaros regulated many anergy-associated genes, including Zfp318, which is implicated in the attenuation of BCR responsiveness by promoting immunoglobulin D expression in anergic B cells. TLR signaling was hyperactive in Ikaros-deficient B cells, which failed to upregulate feedback inhibitors of the MyD88-nuclear factor κB signaling pathway. Systemic inflammation was lost on expression of a non-self-reactive BCR or loss of MyD88 in Ikaros-deficient B cells. Thus, Ikaros acts as a guardian preventing autoimmunity by promoting BCR anergy and restraining TLR signaling.
Sigma-Aldrich
Lysozyme Biotin-Caproyl, lyophilized powder, >20,000 U/mg (Activity of lysozyme used for congugation.)
Pricing and availability is not currently available.
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Pricing and availability is not currently available.