Merck
  • Expression of a CARD Slows the Retinal Degeneration of a Geographic Atrophy Mouse Model.
Expression of a CARD Slows the Retinal Degeneration of a Geographic Atrophy Mouse Model.
Molecular therapy. Methods & clinical development (2019-07-25)
Brianna M Young, Kyle Jones, Michael T Massengill, Erin Walsh, Hong Li, Alfred S Lewin, Cristhian J Ildefonso
ABSTRACT
Age-related macular degeneration (AMD) has been linked to oxidative damage and para-inflammation, an activation of inflammasome signaling in the retinal pigment epithelium (RPE) and the underlying choriocapillaris. Herein, we tested the efficacy of a gene-delivered caspase-1 inhibitor in controlling the retinal degeneration observed in two models of RPE-choroid oxidative damage. In an acute model of oxidative stress (NaIO3 injection), eyes pre-treated with the sGFP-TatCARD (trans-activator of transcription; caspase activation and recruitment domain) vector demonstrated a recovery of retinal function and partial protection of RPE structure 1 month after damage, in contrast with control-treated eyes. In a model of chronic oxidative stress (RPE-specific deletion of Sod2), eyes treated with the sGFP-TatCARD vector after the onset of degeneration had a significantly slower decline in retinal function when compared to control-treated eyes. Earlier treatment of this model with the same adeno-associated virus (AAV) vector resulted in a greater protection of RPE function in eyes treated with the TatCARD when compared to control-treated eyes. Our results demonstrate that intravitreal delivery of sGFP-TatCARD reduces inflammation and can protect the retina from both acute and sustained oxidative damage within the RPE and choroid. Therefore, gene therapy with a cell-penetrating inflammasome inhibitor such as CARD may stem the progression of AMD.
Sigma-Aldrich
Sodium iodate, ≥99%
Sigma-Aldrich
MISSION® esiRNA, targeting human SOD2
Millipore
MILLIPLEX® Mouse Cytokine/Chemokine Magnetic Bead Panel - Immunology Multiplex Assay, Simultaneously analyze multiple cytokine and chemokine biomarkers with Bead-Based Multiplex Assays using the Luminex technology, in mouse serum, plasma and cell culture samples.