Merck
  • Partial decortication ameliorates dopamine depletion‑induced striatal neuron lesions in rats.
Partial decortication ameliorates dopamine depletion‑induced striatal neuron lesions in rats.
International journal of molecular medicine (2019-08-01)
Yaofeng Zhu, Bingbing Liu, Xuefeng Zheng, Jiajia Wu, Si Chen, Zhi Chen, Tao Chen, Ziyun Huang, Wanlong Lei
ABSTRACT
The balance between glutamate (cortex and thalamus) and dopamine (substantia nigra) inputs on striatal neurons is of vital importance. Dopamine deficiency, which breaks this balance and leads to the domination of cortical glutamatergic inputs, plays an important role in Parkinson's disease (PD). However, the exact impact on striatal neurons has not been fully clarified. Thus, the present study aimed to characterize the influence of corticostriatal glutamatergic inputs on striatal neurons after decortication due to dopamine depletion in rats. 6‑Hydroxydopamine was injected into the right medial forebrain bundle to induce dopamine depletion, and/or ibotenic acid into the primary motor cortex to induce decortication. Subsequently, the grip strength test and Morris water maze task indicated that decortication significantly shortened the hang time and the latency that had been increased in the rats subjected to dopamine depletion. Golgi staining and electron microscopy analysis showed that the total dendritic length and dendritic spine density of the striatal neurons were decreased in the dopamine‑depleted rats, whereas decortication alleviated this damage. Immunohistochemistry analysis demonstrated that decortication decreased the number of caspase‑3‑positive neurons in the dopamine‑depleted rats. Moreover, reverse transcription‑quantitative PCR and western blot analyses showed that decortication offset the upregulation of caspase‑3 at both the protein and mRNA levels in the dopamine‑depleted rats. In conclusion, the present study demonstrated that a relative excess of cortical glutamate inputs had a substantial impact on the pathological processes of striatal neuron lesions in PD.
Sigma-Aldrich
Anti-Tyrosine Hydroxylase Antibody, clone LNC1, ascites fluid, clone LNC1, Chemicon®
Sigma-Aldrich
Peroxidase Anti-Peroxidase Soluble Complex antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Mouse IgG (Fc specific) antibody produced in goat, 2.0 mg/mL, affinity isolated antibody
Sigma-Aldrich
Goat Anti-Rabbit IgG Antibody, (H+L) HRP conjugate, 1 mg/mL, Chemicon®