Merck
  • Expression pattern differences between osteoarthritic chondrocytes and mesenchymal stem cells during chondrogenic differentiation.
Expression pattern differences between osteoarthritic chondrocytes and mesenchymal stem cells during chondrogenic differentiation.
Osteoarthritis and cartilage (2010-10-05)
P Bernstein, C Sticht, A Jacobi, C Liebers, S Manthey, M Stiehler
ABSTRACT
The use of mesenchymal stem cells (MSCs) for cartilage regeneration is hampered by lack of knowledge about the underlying molecular differences between chondrogenically stimulated chondrocytes and MSCs. The aim of this study was to evaluate differences in phenotype and gene expression between primary human chondrocytes and MSCs during chondrogenic differentiation in three-dimensional (3D) pellet culture (PC). Chondrocytes isolated from cartilage samples obtained during total knee alloarthroplastic procedure (N=8) and MSCs, purified from bone marrow aspirates of healthy donors (N=8), were cultivated in PC under chondrogenic conditions. Immunohistology and quantitative reverse transcribing PCR (RT-PCR) were performed for chondrogenic-specific markers (i.e., Sox9, Collagen II). Global gene expression of the so-cultivated chondrocytes and MSCs was assessed by a novel approach of microarray-based pathway analysis. Refinement of data was done by hypothesis-driven gene expression omnibus (GEO) dataset comparison. Validation was performed with separate samples in transforming growth factor (TGF)β+ or TGFβ- conditions by use of quantitative real-time RT-PCR. Chondrogenic commitment of both cell types was observed. Interestingly, chondrocytes demonstrated an upregulated fatty acid/cholesterol metabolism which may give hints for future optimization of culture conditions. The novel microarray-based pathway analysis applied in this study seems suitable for the evaluation of whole-genome based array datasets in case when hypotheses can be backed with already existing GEO datasets. Future experiments should further explore the different metabolic behaviour of chondrocytes and MSC.
Sigma-Aldrich
Anti-Sox9 Antibody, Chemicon®, from rabbit
Pricing and availability is not currently available.