Merck
  • Prelamin A acts to accelerate smooth muscle cell senescence and is a novel biomarker of human vascular aging.
Prelamin A acts to accelerate smooth muscle cell senescence and is a novel biomarker of human vascular aging.
Circulation (2010-05-12)
Cassandra D Ragnauth, Derek T Warren, Yiwen Liu, Rosamund McNair, Tamara Tajsic, Nichola Figg, Rukshana Shroff, Jeremy Skepper, Catherine M Shanahan
ABSTRACT
Hutchinson-Gilford progeria syndrome is a rare inherited disorder of premature aging caused by mutations in LMNA or Zmpste24 that disrupt nuclear lamin A processing, leading to the accumulation of prelamin A. Patients develop severe premature arteriosclerosis characterized by vascular smooth muscle cell (VSMC) calcification and attrition. To determine whether defective lamin A processing is associated with vascular aging in the normal population, we examined the profile of lamin A expression in normal and aged VSMCs. In vitro, aged VSMCs rapidly accumulated prelamin A coincidently with nuclear morphology defects, and these defects were reversible by treatment with farnesylation inhibitors and statins. In human arteries, prelamin A accumulation was not observed in young healthy vessels but was prevalent in medial VSMCs from aged individuals and in atherosclerotic lesions, where it often colocalized with senescent and degenerate VSMCs. Prelamin A accumulation correlated with downregulation of the lamin A processing enzyme Zmpste24/FACE1, and FACE1 mRNA and protein levels were reduced in response to oxidative stress. Small interfering RNA knockdown of FACE1 reiterated the prelamin A-induced nuclear morphology defects characteristic of aged VSMCs, and overexpression of prelamin A accelerated VSMC senescence. We show that prelamin A acts to disrupt mitosis and induce DNA damage in VSMCs, leading to mitotic failure, genomic instability, and premature senescence. This study shows that prelamin A is a novel biomarker of VSMC aging and disease that acts to accelerate senescence. It therefore represents a novel target to ameliorate the effects of age-induced vascular dysfunction.
Sigma-Aldrich
Anti-Goat/Sheep IgG−Peroxidase antibody, Mouse monoclonal, clone GT-34, purified from hybridoma cell culture
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)