Merck
  • Genetic changes and histopathological types in colorectal tumors from patients with familial adenomatous polyposis.
Genetic changes and histopathological types in colorectal tumors from patients with familial adenomatous polyposis.
Cancer research (1990-11-15)
M Miyaki, M Seki, M Okamoto, A Yamanaka, Y Maeda, K Tanaka, R Kikuchi, T Iwama, T Ikeuchi, A Tonomura
ABSTRACT
Loss of heterozygosity (LOH) and K-ras mutation were analyzed in 111 colorectal polyps and 26 invasive carcinomas from 40 patients with familial adenomatous polyposis of distinct histopathological types. LOH, being less than 2% in moderate adenomas, was detected on chromosome 5q (20%) in severe adenomas, on 5q (26%) and 17p (38%) in intramucosal carcinomas, and on 5q (52%), 17p (56%), 18 (46%), and 22q (33%) in invasive carcinomas. LOH on chromosome 5q occurred most frequently in the region close to the APC gene both in adenomas and carcinomas, and a loss of the normal allele of the APC gene was demonstrated in 3 cases. K-ras mutation markedly increased in the step of development from moderate (11%) to severe (36%) adenomas. These results suggest the following mechanisms for the development of colon tumors in patients with familial adenomatous polyposis: (a) the heterozygous mutant/wild-type condition at the APC gene causes formation of mild or moderate adenoma; (b) the loss of the normal allele in the APC gene leads to a change from moderate to severe adenoma; (c) LOH on chromosome 17p contributes to the conversion of adenoma to intramucosal carcinoma; (d) LOH on other chromosomes, such as 18 and 22q, are involved in the progression of intramucosal carcinoma to invasive carcinoma; and (e) K-ras mutation may also affect the development of moderate to severe adenoma.