Merck
  • Pyrethroid pesticide metabolite, 3-PBA, in soils: method development and application to real agricultural soils.
Pyrethroid pesticide metabolite, 3-PBA, in soils: method development and application to real agricultural soils.
Environmental science and pollution research international (2018-12-07)
Idalina Bragança, Paulo C Lemos, Cristina Delerue-Matos, Valentina F Domingues
ABSTRACT
3-Phenoxybenzoic acid (3-PBA) is a shared metabolite of several synthetic pyrethroid pesticides (SPs) resulting from environmental degradation of parent compounds and thus occurs frequently as a residue in samples. Hence, the importance of 3-PBA evaluation after pyrethroid application. There is a gap of analytical methods to determine 3-PBA in soil samples. Therefore, an analytical method that combines the solid-phase extraction (SPE) and gas chromatography-mass spectrometry (GC/MS) detection has been developed for the determination of 3-PBA in soil samples. The analytical method was validated in terms of linearity, sensitivity, intra- and inter-day batch precisions, recoveries, and quantification limits. An SPE method using a Strata X cartridge allows obtaining limits of detection and quantification equal to 4.0 and 13.3 ng g-1, respectively. Under optimized conditions, the method average recovery levels ranged from 70.3 to 93.5% with a relative standard deviation below 3.4%. Method intra- and inter-day precision was under 5.0 and 4.8%, respectively. The developed method was applied to 11 agricultural soil samples in the north of Portugal. The developed methodology allowed for the determination of the pyrethroid metabolite, 3-PBA, in agricultural soil samples at levels of few ng g-1. Graphical abstract ᅟ.