Merck
  • Overexpression of ERBB4 rejuvenates aged mesenchymal stem cells and enhances angiogenesis via PI3K/AKT and MAPK/ERK pathways.
Overexpression of ERBB4 rejuvenates aged mesenchymal stem cells and enhances angiogenesis via PI3K/AKT and MAPK/ERK pathways.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2018-12-20)
Xiaoting Liang, Yue Ding, Fang Lin, Yuelin Zhang, Xiaohui Zhou, Qingshu Meng, Xingyue Lu, Guojun Jiang, Hongming Zhu, Yu Chen, Qizhou Lian, Huimin Fan, Zhongmin Liu
ABSTRACT
The age-related functional exhaustion limits potential efficacy of mesenchymal stem cells (MSC) in treating cardiovascular disease. Therefore, rejuvenation of aged MSC in the elderly population is of great interest. We have previously reported that Erb-B2 receptor tyrosine kinase 4 ( ERBB4) plays a critical role in regulating MSC survival under hypoxia. The aim of this study was to investigate whether ERBB4 rejuvenates aged MSC and how ERBB4 enhances therapeutic efficacy of aged MSC in treating myocardial infarction (MI). Compared with vector aged MSC (aged-MSC), ERBB4-engineered aged MSC (ER4-aged-MSC) conferred resistance to oxidative stress-induced cell death and ameliorated the senescent phenotype in vitro. Four weeks after MI, the ER4-aged-MSC group exhibited enhanced blood vessel density, reduced cardiac remodeling and apoptosis with improved heart function compared with the aged-MSC group. Overexpression of ERBB4 caused an increase in phosphorylated v-akt murine thymoma viral oncogene homolog 1 (AKT), and phosphorylated ERK expression under hypoxia. ER4-aged-MSC secreted higher levels of angiopoietin, epithelial neutrophil activating peptide 78, VEGF, and fibroblast growth factor 2, and enhanced tube formation in HUVEC. The impact of ERBB4 on protein expression, proangiogenesis, cell behavior, and cytokine secretion was abolished by inhibiting PI3K/AKT and MAPK/ERK signaling pathway.-Liang, X., Ding, Y., Lin, F., Zhang, Y., Zhou, X., Meng, Q., Lu, X., Jiang, G., Zhu, H., Chen, Y., Lian, Q., Fan, H., Liu, Z. Overexpression of ERBB4 rejuvenates aged mesenchymal stem cells and enhances angiogenesis via PI3K/AKT and MAPK/ERK pathways.