Merck
  • Oxidative Stress of Carbon Nanotubes on Proteins Is Mediated by Metals Originating from the Catalyst Remains.
Oxidative Stress of Carbon Nanotubes on Proteins Is Mediated by Metals Originating from the Catalyst Remains.
ACS nano (2019-01-27)
Atsushi Hirano, Momoyo Wada, Takeshi Tanaka, Hiromichi Kataura
ABSTRACT
Nanomaterials introduced into biological systems are immediately coated by proteins in vivo. They induce oxidative stress on adsorbed proteins and hence alter the protein structures, which determines the fate pathways and biological impacts of nanomaterials. Carbon nanotubes (CNTs) have been suggested to cause protein oxidation. In this work, we discovered that CNTs induce oxidative stress on proteins in cooperation with coexisting metals originating from catalyst remains. Protein sulfhydryl groups were readily oxidized by the coexistence of CNTs and metals. Numerical simulations of the reaction demonstrated that the metals effectively mediate electron transfer between the CNTs and protein sulfhydryl groups. Thus, the coexistence of CNTs and metals, even in low concentrations, generates oxidative stress on proteins with high reaction rates. Metal catalysts used for CNT growth, in turn, catalyze the oxidation reaction of proteins. The proposed protein oxidation mechanism will advance the fundamental understanding of the biological safety and toxicity of nanomaterials synthesized using metal catalysts.