Merck
  • An evaluation of the effect of incorporating metal salts into 1,8 diazafluoren-9-one (DFO) formulations for fingermark enhancement.
An evaluation of the effect of incorporating metal salts into 1,8 diazafluoren-9-one (DFO) formulations for fingermark enhancement.
Science & justice : journal of the Forensic Science Society (2019-05-06)
Kelly Mayse, Vaughn G Sears, Niko Nicolasora, Stephen Bleay
ABSTRACT
A study into the modification of 1,8-diazafluoren-9-one (DFO) formulations by the additions of metal salts into the working solution is reported. Similar additions have been found to increase the fluorescence of marks developed using other amino acid reagents including 1,2-indandione and the ninhydrin analogue 5-methylthioninhydrin. It was found that adding zinc chloride to give a 1:1 ratio of zinc ions:DFO molecules gave optimum fluorescence, and improvements in performance over the standard DFO formulation were achieved. Attempts to produce equivalent formulations with iron, nickel and palladium chlorides were unsuccessful. In a comparative trial with a 1,2-indandione-zinc formulation on brown paper and cardboard substrates, 1,2-indandione-zinc gave superior results and it was decided to focus further research on this reagent instead of DFO-zinc.