Merck
  • Deficiency of Socs3 leads to brain-targeted EAE via enhanced neutrophil activation and ROS production.
Deficiency of Socs3 leads to brain-targeted EAE via enhanced neutrophil activation and ROS production.
JCI insight (2019-04-03)
Zhaoqi Yan, Wei Yang, Luke Parkitny, Sara A Gibson, Kevin S Lee, Forrest Collins, Jessy S Deshane, Wayne Cheng, Amy S Weinmann, Hairong Wei, Hongwei Qin, Etty N Benveniste
ABSTRACT
Dysregulation of the JAK/STAT signaling pathway is associated with Multiple Sclerosis (MS) and its mouse model, Experimental Autoimmune Encephalomyelitis (EAE). Suppressors Of Cytokine Signaling (SOCS) negatively regulate the JAK/STAT pathway. We previously reported a severe, brain-targeted, atypical form of EAE in mice lacking Socs3 in myeloid cells (Socs3ΔLysM), which is associated with cerebellar neutrophil infiltration. There is emerging evidence that neutrophils are detrimental in the pathology of MS/EAE, however, their exact function is unclear. Here we demonstrate that neutrophils from the cerebellum of Socs3ΔLysM mice show a hyper-activated phenotype with excessive production of reactive oxygen species (ROS) at the peak of EAE. Neutralization of ROS in vivo delayed the onset and reduced severity of atypical EAE. Mechanistically, Socs3-deficient neutrophils exhibit enhanced STAT3 activation, a hyper-activated phenotype in response to G-CSF, and upon G-CSF priming, increased ROS production. Neutralization of G-CSF in vivo significantly reduced the incidence and severity of the atypical EAE phenotype. Overall, our work elucidates that hypersensitivity of G-CSF/STAT3 signaling in Socs3ΔLysM mice leads to atypical EAE by enhanced neutrophil activation and increased oxidative stress, which may explain the detrimental role of G-CSF in MS patients.
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC)