Merck
  • Polyamines and eIF5A Hypusination Modulate Mitochondrial Respiration and Macrophage Activation.
Polyamines and eIF5A Hypusination Modulate Mitochondrial Respiration and Macrophage Activation.
Cell metabolism (2019-05-28)
Daniel J Puleston, Michael D Buck, Ramon I Klein Geltink, Ryan L Kyle, George Caputa, David O'Sullivan, Alanna M Cameron, Angela Castoldi, Yaarub Musa, Agnieszka M Kabat, Ying Zhang, Lea J Flachsmann, Cameron S Field, Annette E Patterson, Stefanie Scherer, Francesca Alfei, Francesc Baixauli, S Kyle Austin, Beth Kelly, Mai Matsushita, Jonathan D Curtis, Katarzyna M Grzes, Matteo Villa, Mauro Corrado, David E Sanin, Jing Qiu, Nora Pällman, Katelyn Paz, Maria Elena Maccari, Bruce R Blazar, Gerhard Mittler, Joerg M Buescher, Dietmar Zehn, Sabine Rospert, Edward J Pearce, Stefan Balabanov, Erika L Pearce
ABSTRACT
How cells adapt metabolism to meet demands is an active area of interest across biology. Among a broad range of functions, the polyamine spermidine is needed to hypusinate the translation factor eukaryotic initiation factor 5A (eIF5A). We show here that hypusinated eIF5A (eIF5AH) promotes the efficient expression of a subset of mitochondrial proteins involved in the TCA cycle and oxidative phosphorylation (OXPHOS). Several of these proteins have mitochondrial targeting sequences (MTSs) that in part confer an increased dependency on eIF5AH. In macrophages, metabolic switching between OXPHOS and glycolysis supports divergent functional fates stimulated by activation signals. In these cells, hypusination of eIF5A appears to be dynamically regulated after activation. Using in vivo and in vitro models, we show that acute inhibition of this pathway blunts OXPHOS-dependent alternative activation, while leaving aerobic glycolysis-dependent classical activation intact. These results might have implications for therapeutically controlling macrophage activation by targeting the polyamine-eIF5A-hypusine axis.
Sigma-Aldrich
Oligomycin from Streptomyces diastatochromogenes, ≥90% total oligomycins basis (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
IPTG, ≥99% (TLC), ≤0.1% Dioxane
Pricing and availability is not currently available.
Sigma-Aldrich
Antimycin A from Streptomyces sp.
Pricing and availability is not currently available.
Sigma-Aldrich
Rotenone, ≥95%
Pricing and availability is not currently available.
Sigma-Aldrich
Tamoxifen, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-LC3B antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Pricing and availability is not currently available.
Sigma-Aldrich
Ciclopirox, ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Carbonyl cyanide 4-(trifluoromethoxy)phenylhydrazone, ≥98% (TLC), powder
Pricing and availability is not currently available.
Sigma-Aldrich
Deoxyhypusine Synthase Inhibitor, GC7, The Deoxyhypusine Synthase Inhibitor, GC7, also referenced under CAS 150333-69-0, controls the biological activity of Deoxyhypusine Synthase.
Pricing and availability is not currently available.