Merck
Breast cancer cell motility is promoted by 14-3-3γ.
Breast cancer (Tokyo, Japan) (2019-03-05)
Emiko Hiraoka, Takahiro Mimae, Masaoki Ito, Takayuki Kadoya, Yoshihiro Miyata, Akihiko Ito, Morihito Okada
ABSTRACT
Pseudopodia are actin-rich ventral protrusions associated with cell motility and cancer cell invasion. We previously applied our established method of using excimer laser cell etching to isolate pseudopodial proteins from MDA-MB-231 breast cancer cells. We later identified 14-3-3γ as an oncogenic molecule among 46 candidate proteins that are specific to pseudopodia. The present study aimed to determine the function of 14-3-3γ in the motility of breast cancer cells. MDA-MB-231 cells were cultured on 3-µm porous membranes and double stained to localize 14-3-3γ and phalloidin in pseudopodia using confocal imaging. We assessed pseudopodia numbers and length, as well as migration and wound healing in MDA-MB-231 cells with knockdown and forced expression of 14-3-3γ to determine 14-3-3γ involvement in cell motility. We also immunohistochemically analyzed 14-3-3γ in human breast cancer tissues with high-grade lymphatic invasion. We specifically located 14-3-3γ in pseudopodia of MDA-MB-231 cells. Knockdown and forced expression of 14-3-3γ, respectively, decreased and increased pseudopodial formation and elongation. Migration and wound healing assays also showed that 14-3-3γ knockdown and forced expression, respectively, decreased and increased the number of underside cells and acellular areas in MDA-MB-231 breast cancer cells. More 14-3-3γ was expressed in sites of lymphatic invasion, than in the center and periphery of human breast cancer tissues. The role of 14-3-3γ in breast cancer invasiveness might be to promote cell motility. Inhibition of 14-3-3γ could, therefore, become a novel target of therapy to prevent invasion and metastasis in patients with breast cancers expressing 14-3-3γ.