- Crystal structures, spectroscopic properties of new cobalt(II), nickel(II), zinc(II) and palladium(II) complexes derived from 2-acetyl-5-chloro thiophene thiosemicarbazone: Anticancer evaluation.
Crystal structures, spectroscopic properties of new cobalt(II), nickel(II), zinc(II) and palladium(II) complexes derived from 2-acetyl-5-chloro thiophene thiosemicarbazone: Anticancer evaluation.
The reactions of cobalt(II), nickel(II), zinc(II) chlorides and [Pd(DMSO)2Cl2] with 2-acetyl-5-chloro-thiophene thiosemicarbazone (HL) leads to the formation of a series of new complexes: [CoCl2(S-HL)2], 1; [Ni(N,S-L)2], 2 [ZnCl2(S-HL)2], 3 and [PdCl2(N,S-HL)], 4. All the complexes have been characterized by elemental analysis, IR, LC-MS. 1H and 13C NMR spectroscopy have been performed for Zn(II) and Pd(II) complexes. The crystal structures of the complexes 1-3 have been determined by single-crystal X-ray diffraction methods. The compounds, (1) and (3), crystallized in the monoclinic crystal system with C2/c space group. In both complexes, the metal centers are four-coordinated in a distorted tetrahedral configuration by two sulfur atoms from two thiosemicarbazone ligands and two Cl anions. The crystal structure of (2) consists of monomeric entities where the nickel(II) ion exhibit distorted square planar geometry. The coordination geometry around nickel ion is four-coordinate with four atoms of the two chelating thiosemicarbazone ligands which are in cis position. The τ4 value of 0.255 obtained from the τ4 analysis of complex (2) shows that the four-coordinate geometry around the central nickel ion is close to square planar. Complex (4) is mononuclear, the central ion is coordinated through the sulfur and the azomethine nitrogen atom of neutral ligand. The cytotoxic effects of all complexes were analyzed for three cancer cell lines, Caco-2, DLD-1, and SW620 compared to normal colon epithelial cell line, CCD18Co. Complex (4) is more active against DLD-1, SW620 and Caco-2 than CCD18Co. The efficiency of complex (4) is more effective in aggressive cancer cell lines. Therefore, it can be used as a new chemotherapeutic, especially in the treatment of resistant cancer types caused by long-term use of platinum-based drugs.