Merck
  • Long-term Effects of Aripiprazole Treatment during Adolescence on Cognitive Function and Dopamine D2 Receptor Expression in Neurodevelopmentally Normal Rats.
Long-term Effects of Aripiprazole Treatment during Adolescence on Cognitive Function and Dopamine D2 Receptor Expression in Neurodevelopmentally Normal Rats.
Clinical psychopharmacology and neuroscience : the official scientific journal of the Korean College of Neuropsychopharmacology (2019-07-30)
Hyung Jun Choi, Soo Jung Im, Hae Ri Park, Subin Park, Chul-Eung Kim, Seunghyong Ryu
ABSTRACT
This study aimed to investigate the long-term effects of aripiprazole treatment during adolescence on behavior, cognitive function, and dopamine D2 receptor (D2R) expression in adult rats. Adolescent male Sprague-Dawley rats were injected intraperitoneally with aripiprazole, risperidone, or vehicle control for 3 weeks (postnatal day 36‒56). After a 2-week washout period, locomotion, anxiety, and spatial working memory were evaluated in adulthood (postnatal day 71‒84), using an open field test, elevated plus maze, and Y-maze, respectively. In addition, we assessed D2R levels in the dorsolateral and medial prefrontal cortex (PFC), dorsal and ventral striatum, and hippocampus using western blot analysis. Spontaneous alternation performance (SAP) in the Y-maze, a measure of spatial working memory, differed significantly among the 3 groups (F = 3.89, p = 0.033). A post-hoc test confirmed that SAP in the aripiprazole group was significantly higher than that in the risperidone group ( post-hoc test p = 0.013). D2R levels in the medial PFC (F= 8.72, p= 0.001) and hippocampus (F= 13.54, p > 0.001) were different among the 3 groups. D2R levels in the medial PFC and hippocampus were significantly lower in the aripiprazole-treated rats than that in the risperidone-treated rats (post-hoc test p = 0.025 and p > 0.001, respectively) and controls (post-hoc test p > 0.001, all). This study showed that aripiprazole treatment in adolescence could influence cognitive function and dopaminergic neurotransmission into early adulthood.