MilliporeSigma
  • Anti-platelet action of isoliquiritigenin, an aldose reductase inhibitor in licorice.
Anti-platelet action of isoliquiritigenin, an aldose reductase inhibitor in licorice.
European journal of pharmacology (1992-02-25)
M Tawata, K Aida, T Noguchi, Y Ozaki, S Kume, H Sasaki, M Chin, T Onaya
ABSTRACT
The mechanism was studied by which isoliquiritigenin, a new aldose reductase inhibitor purified from licorice (Glycyrrhizae radix), inhibits platelet aggregation. This new agent significantly inhibited the phosphorylation of 40,000- and 20,000-dalton proteins, and inhibited the formation of 12 (S)-hydroxy-5,8,10-heptadecatrienoic acid, 12-hydroxyeicosatetraenoic acid and thromboxane B2. The inhibitory effect of isoliquiritigenin on platelet aggregation in vitro was comparable to that of aspirin. Our findings may indicate that isoliquiritigenin elicits an anti-platelet action by inhibiting not only cyclooxygenase but also lipoxygenase or peroxidase activity in platelets. Isoliquiritigenin also showed an anti-platelet action in vivo. Isoliquiritigenin appears to be the only aldose reductase inhibitor with a significant anti-platelet action. Since the hyperaggregability of platelets has been implicated in the pathogenesis of diabetic complications, isoliquiritigenin may offer a unique benefit as an aldose reductase inhibitor.
Sigma-Aldrich
Isoliquiritigenin, powder