- Acidity and Cd2+ fluorescent sensing and selective CO2 adsorption by a water-stable Eu-MOF.
Acidity and Cd2+ fluorescent sensing and selective CO2 adsorption by a water-stable Eu-MOF.
Dalton transactions (Cambridge, England : 2003) (2019-03-13)
Wei-Ming Liao, Mei-Juan Wei, Jun-Ting Mo, Peng-Yan Fu, Ya-Nan Fan, Mei Pan, Cheng-Yong Su
PMID30860246
ABSTRACT
A new Eu-MOF was designed from an amino-functionalized ligand and Eu(iii) ions under solvothermal conditions. It is a highly porous, water-stable, and luminescent material, exhibiting pH sensing in the acidic range of pH = 7-3 with selective detection for Cd2+ by an enhanced fluorescence of ∼23-fold against a series of metal ions. Gas adsorption performance shows that Eu-MOF exhibits a high CO2/N2 (15/85) selectivity of 109.4 at 273 K and 1 atm. This performance was superior to that of most reported Ln-MOFs owing to its appropriate pore size, dipole-dipole interaction and donor-acceptor Lewis affinities.