Merck
  • A Versatile Protocol for Studying Anterior Cruciate Ligament Reconstruction in a Rabbit Model.
A Versatile Protocol for Studying Anterior Cruciate Ligament Reconstruction in a Rabbit Model.
Tissue engineering. Part C, Methods (2019-03-20)
Mario Hevesi, João F Crispim, Carlo A Paggi, Amel Dudakovic, Wouter van Genechten, Timothy Hewett, Sanjeev Kakar, Aaron J Krych, Andre J van Wijnen, Daniel B F Saris
ABSTRACT
Anterior cruciate ligament (ACL) injuries are frequent, as >200,000 injuries occur in the United States alone each year. Owing to the risks for associated meniscus and cartilage damage, ACL injuries are a significant source of both orthopedic care and research. Given the extended recovery course after ACL injury, which often lasts 1-2 years, and is associated with limited participation in sports and activities of daily living for patients, there is a critical need for the evolution of new and improved methods for ACL repair. Subsequently, animal models of ACL reconstruction (ACLR) play a key role in the development and initial trialing of novel ACL interventions. This article provides a clear operative description and associated illustrations for a validated, institutional animal care and use committee, and veterinarian approved and facile model of ACLR to serve researchers investigating ACLR.