- Polysialic acid masks neural cell adhesion molecule antigenicity.
Polysialic acid masks neural cell adhesion molecule antigenicity.
The neural cell adhesion molecule (NCAM) is a transmembrane protein involved in major cellular processes. The addition of polysialic acid (PSA), a post-translational modification (PTM) almost exclusively carried by NCAM, alters NCAM properties and functions and is therefore tightly regulated. Changes in NCAM and PSA-NCAM take place during development and ageing and occur in various diseases. The presence of PTMs can reduce the accessibility of antibodies to their epitopes and lead to false negative results. Thus, it is vital to identify antibodies that can specifically detect their target regardless of the presence of PTMs. In the present study, four commercially available NCAM antibodies were characterized by western blot and immunocytochemistry. Antibody specificity was determined by decreasing NCAM expression with small interfering RNA and subsequently determining whether the antibodies still produced a signal. In addition, PSA was digested with endoneuraminidase N to assess whether removing PSA improves NCAM detection with these antibodies. Our study revealed that the presence of PSA on NCAM reduced antibody accessibility to the epitope and consequently masked NCAM antigenicity for both techniques investigated. Moreover, three of the four antibodies tested were specific for the detection of NCAM by western blot and by immunocytochemistry. Altogether, this study demonstrates the importance of choosing the correct antibody to study NCAM depending on the technique of interest and underlines the importance of taking PTMs into account when using antibody-based techniques for the study of NCAM.