Merck
  • Blood autophagy defect causes accelerated non-hematopoietic organ aging.
Blood autophagy defect causes accelerated non-hematopoietic organ aging.
Aging (2019-07-23)
Yixuan Fang, Lingjiang Zhu, Ni An, Gaoyue Jiang, Jiawei Qian, Ruijin Zhao, Na Yuan, Suping Zhang, Jianrong Wang
ABSTRACT
Autophagy has been well studied in regulating aging; however, the impact of autophagy in one organ on the aging of other organs has not been documented. In this study, we used a mouse model with deletion of an autophagy-essential gene Atg7 in hematopoietic system to evaluate the intrinsic role of hematopoietic autophagy on the aging of non-hematopoietic organs. We found that autophagy defect in hematopoietic system causes growth retardation and shortened lifespan, along with aging-like phenotypes including hypertrophic heart, lung and spleen, but atrophic thymus and reduced bone mineral density at organismal level. Hematopoietic autophagy defect also causes increased oxidative stress and mitochondrial mass or aging gene expression at cellular level in multiple non-hematopoietic organs. The organ aging in the Atg7-deleted mice was reversed by anatomic connection to wild-type mice with intact blood autophagy via parabiosis, but not by injection of blood cell-free plasma. Our finding thus highlights an essential role of hematopoietic autophagy for decelerating aging in non-hematopoietic organs.