Merck
Polyelectrolyte-Stabilised Magnetic-Plasmonic Nanocomposites.
Nanomaterials (Basel, Switzerland) (2018-12-16)
Shelley Stafford, Coralie Garnier, Yurii K Gun'ko
ABSTRACT
In this work, new magnetic-plasmonic nanocomposites have been developed through the use of two complementary polyelectrolytes⁻polystyrene sulfonate (PSS) and poly(allylamine hydrochloride) (PAH). PSS, a negatively charged polyelectrolyte, was utilized as a stabiliser for magnetite nanoparticles, and PAH, a positively charged polyelectrolyte, was used to stabilize gold nanoparticles. The combination of these two entities resulted in a magnetic-plasmonic nanocomposite that is highly reproducible and scalable. This approach was found to work for a variety of PSS concentrations. The produced magnetic-plasmonic nanomaterials have been characterized by vibrational sample magnetometry (VSM), transmission electron microscopy (TEM) and UV-Vis spectroscopy. These nanocomposite materials have the potential to be used in a variety of biological applications including bioseparation and biosensing.