Merck
  • Tie2 activation promotes choriocapillary regeneration for alleviating neovascular age-related macular degeneration.
Tie2 activation promotes choriocapillary regeneration for alleviating neovascular age-related macular degeneration.
Science advances (2019-02-23)
Jaeryung Kim, Jang Ryul Park, Jeongwoon Choi, Intae Park, Yoonha Hwang, Hosung Bae, Yongjoo Kim, WooJhon Choi, Jee Myung Yang, Sangyeul Han, Tae-Young Chung, Pilhan Kim, Yoshiaki Kubota, Hellmut G Augustin, Wang-Yuhl Oh, Gou Young Koh
ABSTRACT
Choriocapillary loss is a major cause of neovascular age-related macular degeneration (NV-AMD). Although vascular endothelial growth factor (VEGF) blockade for NV-AMD has shown beneficial outcomes, unmet medical needs for patients refractory or tachyphylactic to anti-VEGF therapy exist. In addition, the treatment could exacerbate choriocapillary rarefaction, necessitating advanced treatment for fundamental recovery from NV-AMD. In this study, Tie2 activation by angiopoietin-2-binding and Tie2-activating antibody (ABTAA) presents a therapeutic strategy for NV-AMD. Conditional Tie2 deletion impeded choriocapillary maintenance, rendering eyes susceptible to NV-AMD development. Moreover, in a NV-AMD mouse model, ABTAA not only suppressed choroidal neovascularization (CNV) and vascular leakage but also regenerated the choriocapillaris and relieved hypoxia. Conversely, VEGF blockade degenerated the choriocapillaris and exacerbated hypoxia, although it suppressed CNV and vascular leakage. Together, we establish that angiopoietin-Tie2 signaling is critical for choriocapillary maintenance and that ABTAA represents an alternative, combinative therapeutic strategy for NV-AMD by alleviating anti-VEGF adverse effects.
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
Anti-Green Fluorescent Protein Antibody, Chemicon®, from chicken
Sigma-Aldrich
Anti-Opsin Antibody, Red/Green, Chemicon®, from rabbit
Sigma-Aldrich
Anti-PECAM-1 Antibody, clone 2H8, Azide Free, clone 2H8, Chemicon®, from hamster(Armenian)