Merck
  • Vigorous, but differential mononuclear cell response of cirrhotic patients to bacterial ligands.
Vigorous, but differential mononuclear cell response of cirrhotic patients to bacterial ligands.
World journal of gastroenterology (2011-04-02)
Varenka J Barbero-Becerra, María Concepción Gutiérrez-Ruiz, Carmen Maldonado-Bernal, Félix I Téllez-Avila, Roberto Alfaro-Lara, Florencia Vargas-Vorácková
ABSTRACT
To study the role of gram-positive and gram-negative bacteria in the pathogenesis of liver injury, specifically the activation of inflammatory mediators. Peripheral blood mononuclear cells of 20 out-patients were studied, 10 of them with cirrhosis. Peripheral blood mononuclear cells were isolated and exposed to lipopolysaccharide or lipoteichoic acid. CD14, Toll-like receptor 2 and 4 expression was determined by flow cytometry, and tumor necrosis factor (TNF) α, interleukin (IL)-1β, IL-6, IL-12 and IL-10 secretion in supernatants was determined by ELISA. Higher CD14, Toll-like receptor 2 and 4 expression was observed in peripheral blood mononuclear cells from cirrhotic patients, (P < 0.01, P < 0.006, P < 0.111) respectively. Lipopolysaccharide and lipoteichoic acid induced a further increase in CD14 expression (P < 0.111 lipopolysaccharide, P < 0.013 lipoteichoic acid), and a decrease in Toll-like receptor 2 (P < 0.008 lipopolysaccharide, P < 0.008 lipoteichoic acid) and Toll-like receptor 4 (P < 0.008 lipopolysaccharide, P < 0.028 lipoteichoic acid) expression. With the exception of TNFα, absolute cytokine secretion of peripheral blood mononuclear cells was lower in cirrhotic patients under non-exposure conditions (P < 0.070 IL-6, P < 0.009 IL-1β, P < 0.022 IL-12). Once exposed to lipopolysaccharide or lipoteichoic acid, absolute cytokine secretion of peripheral blood mononuclear cells was similar in cirrhotic and non-cirrhotic patients, determining a more vigorous response in the former (P < 0.005 TNFα, IL-1β, IL-6, IL-2 and IL-10 lipopolysaccharide; P < 0.037 TNFα; P < 0.006 IL-1β; P < 0.005 IL-6; P < 0.007 IL-12; P < 0.014 IL-10 lipoteichoic acid). Response of peripheral blood mononuclear cells was more intense after lipopolysaccharide than after lipoteichoic acid exposure. Peripheral blood mononuclear cells of cirrhotic patients are able to respond to a sudden bacterial ligand exposure, particularly lipopolysaccharide, suggesting that immune regulation mechanisms are still present.
Sigma-Aldrich
Lipoteichoic acid from Bacillus subtilis