Merck
  • Interleukin-6-type cytokines in neuroprotection and neuromodulation: oncostatin M, but not leukemia inhibitory factor, requires neuronal adenosine A1 receptor function.
Interleukin-6-type cytokines in neuroprotection and neuromodulation: oncostatin M, but not leukemia inhibitory factor, requires neuronal adenosine A1 receptor function.
Journal of neurochemistry (2010-07-06)
Shamsudheen Moidunny, Raquel Baptista Dias, Evelyn Wesseling, Yuko Sekino, Hendrikus W G M Boddeke, Ana Maria Sebastião, Knut Biber
ABSTRACT
Neuroprotection is one of the prominent functions of the interleukin (IL)-6-type cytokine family, for which the underlying mechanism(s) are not fully understood. We have previously shown that neuroprotection and neuromodulation mediated by IL-6 require neuronal adenosine A(1) receptor (A(1) R) function. We now have investigated whether two other IL-6-type cytokines [oncostatin M (OSM) and leukemia inhibitory factor (LIF)] use a similar mechanism. It is presented here that OSM but not LIF, enhanced the expression of A(1) Rs (both mRNA and protein levels) in cultured neurons. Whereas the neuroprotective effect of LIF was unchanged in A(1) R deficient neurons, OSM failed to protect neurons in the absence of A(1) R. In addition, OSM pre-treatment for 4 h potentiated the A(1) R-mediated inhibition of electrically evoked excitatory post-synaptic currents recorded from hippocampal slices either under normal or hypoxic conditions. No such effect was observed after LIF pre-treatment. Our findings thus strongly suggest that, despite known structural and functional similarities, OSM and LIF use different mechanisms to achieve neuroprotection and neuromodulation.