Merck
  • Zinc as a cofactor for heparin neutralization by histidine-rich glycoprotein.
Zinc as a cofactor for heparin neutralization by histidine-rich glycoprotein.
The Journal of biological chemistry (1997-05-23)
B A Kluszynski, C Kim, W P Faulk
ABSTRACT
We have studied the ability of histidine-rich glycoprotein (HRG) to neutralize the anticoagulant activity of heparin in plasma and in a purified component clotting assay. Addition of HRG to plasma or to the purified component assay did not neutralize the anticoagulant activity of heparin unless micromolar concentrations of zinc were present. Higher zinc concentrations were required for citrated than for heparinized plasmas due to competition of citrate with HRG for zinc binding. Zinc concentrations as low as 1.25 microM revealed HRG to be a powerful competitor of antithrombin for heparin in the purified component assays. HRG binding of heparin also was shown by affinity chromatography of HRG from immobilized heparin in the presence and absence of zinc. In the absence of zinc, HRG was eluted by 0.1 M NaCl, but, in the presence of zinc, elution of HRG required 1.0 M NaCl. Investigation of other divalent cations (copper and magnesium) indicated that augmentation of heparin binding by HRG in the presence of antithrombin was restricted to zinc. The HRG.Zn complex effectively competes with antithrombin for heparin, which restricts the availability of heparin to bind antithrombin and allows thrombin-mediated fibrinogenesis to proceed unimpeded. This could be initiated by zinc released from activated platelets.
Sigma-Aldrich
Heparin−Agarose, (1:1 suspension in a 20% ethanol solution)