Merck
Corticosteroid receptors, macrophages and cardiovascular disease.
Journal of molecular endocrinology (2009-01-23)
Amanda J Rickard, Morag J Young
ABSTRACT
The mineralocorticoid receptor (MR) and glucocorticoid receptor are ligand-activated transcription factors that have important physiological and pathophysiological actions in a broad range of cell types including monocytes and macrophages. While the glucocorticoids cortisol and corticosterone have well-described anti-inflammatory actions on both recruited and tissue resident macrophages, a role for the mineralocorticoid aldosterone in these cells is largely undefined. Emerging evidence, however, suggests that MR signalling may promote pro-inflammatory effects. This review will discuss the current understanding of the role of corticosteroid receptors in macrophages and their effect on diseases involving inflammation, with a particular focus on cardiovascular disease.
Sigma-Aldrich
Corticosterone, ≥98.5% (HPLC)