- DNA-binding activity of TNF-alpha inducing protein from Helicobacter pylori.
DNA-binding activity of TNF-alpha inducing protein from Helicobacter pylori.
Tumor necrosis factor-alpha (TNF-alpha) inducing protein (Tipalpha) is a carcinogenic factor secreted from Helicobacter pylori (H. pylori), mediated through both enhanced expression of TNF-alpha and chemokine genes and activation of nuclear factor-kappaB. Since Tipalpha enters gastric cancer cells, the Tipalpha binding molecules in the cells should be investigated. The direct DNA-binding activity of Tipalpha was observed by pull down assay using single- and double-stranded genomic DNA cellulose. The surface plasmon resonance assay, indicating an association between Tipalpha and DNA, revealed that the affinity of Tipalpha for (dGdC)10 is 2400 times stronger than that of del-Tipalpha, an inactive Tipalpha. This suggests a strong correlation between DNA-binding activity and carcinogenic activity of Tipalpha. And the DNA-binding activity of Tipalpha was first demonstrated with a molecule secreted from H. pylori.