Merck
  • Reduced cortical BDNF expression and aberrant memory in Carf knock-out mice.
Reduced cortical BDNF expression and aberrant memory in Carf knock-out mice.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2010-06-04)
Kelli A McDowell, Ashley N Hutchinson, Sarah J E Wong-Goodrich, Matthew M Presby, Dan Su, Ramona M Rodriguiz, Krystal C Law, Christina L Williams, William C Wetsel, Anne E West
ABSTRACT
Transcription factors are a key point of convergence between the cell-intrinsic and extracellular signals that guide synaptic development and brain plasticity. Calcium-response factor (CaRF) is a unique transcription factor first identified as a binding protein for a calcium-response element in the gene encoding brain-derived neurotrophic factor (Bdnf). We have now generated Carf knock-out (KO) mice to characterize the function of this factor in vivo. Intriguingly, Carf KO mice have selectively reduced expression of Bdnf exon IV-containing mRNA transcripts and BDNF protein in the cerebral cortex, whereas BDNF levels in the hippocampus and striatum remain unchanged, implicating CaRF as a brain region-selective regulator of BDNF expression. At the cellular level, Carf KO mice show altered expression of GABAergic proteins at striatal synapses, raising the possibility that CaRF may contribute to aspects of inhibitory synapse development. Carf KO mice show normal spatial learning in the Morris water maze and normal context-dependent fear conditioning. However they have an enhanced ability to find a new platform location on the first day of reversal training in the water maze and they extinguish conditioned fear more slowly than their wild-type littermates. Finally, Carf KO mice show normal short-term (STM) and long-term memory (LTM) in a novel object recognition task, but exhibit impairments during the remote memory phase of testing. Together, these data reveal novel roles for CaRF in the organization and/or function of neural circuits that underlie essential aspects of learning and memory.
Sigma-Aldrich
Anti-GABA A Receptor β 2,3 Chain Antibody, clone BD17, clone BD17, Chemicon®, from mouse
Sigma-Aldrich
Anti-MAP2 Antibody, Chemicon®, from chicken
Sigma-Aldrich
Anti-Actin Antibody, clone C4, ascites fluid, clone C4, Chemicon®
Sigma-Aldrich
Anti-CREB Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-GAD67 Antibody, clone 1G10.2, clone 1G10.2, Chemicon®, from mouse
Sigma-Aldrich
Anti-Post Synaptic Density Protein 95 Antibody, clone 6G6-1C9, clone 6G6-1C9, Chemicon®, from mouse