Merck
  • v-Abl-mediated apoptotic suppression is associated with SHC phosphorylation without concomitant mitogen-activated protein kinase activation.
v-Abl-mediated apoptotic suppression is associated with SHC phosphorylation without concomitant mitogen-activated protein kinase activation.
The Journal of biological chemistry (1995-03-17)
P J Owen-Lynch, A K Wong, A D Whetton
ABSTRACT
A temperature-sensitive mutant of the v-Abl protein has previously been shown to exhibit tyrosine protein kinase activity in Interleukin 3 (IL-3)-dependent IC.DP cells grown at the permissive temperature (32 degrees C) but not at the restrictive temperature (39 degrees C). These IC.DP cells are dependent on IL-3 for suppression of apoptosis at 39 degrees C, but at 32 degrees C cells will survive without added growth factor. Both IL-3 and v-Abl stimulated the tyrosine phosphorylation of SHC and GTPase-activating protein. However, while IL-3 stimulated similar levels of tyrosine phosphorylation in p46shc and p52shc, v-Abl preferentially phosphorylated p52shc, an event that occurred within 1 h of temperature switch. v-Abl also differentially associated with p46shc in a temperature-independent manner. In contrast, only IL-3 stimulated detectable increases in both myelin basic protein kinase and mitogen-activated protein (MAP) kinase kinase in in vitro assays, although in more specific MAP kinase activity assays a very slight increase in the activity of this enzyme was observed after 6 h at the permissive temperature. Time course studies suggest that phosphorylation and association of SHC with v-Abl is insufficient to lead to significant activation of MAP kinase and that activation of the MAP kinase kinase/MAP kinase pathway is not required for apoptotic suppression.